siRNA Details
Gene Symbol: 4930422I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015180.1 | - | 1 | 289 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714731.1 | - | 1 | 289 | Chr. 10 - 115868862 - 115869522 [-] on Build GRCm38 |
