siRNA Details
Gene Symbol: 4930423D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015187.1 | - | 1 | 1502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB533931.1 | - | 1 | 149 | Chr. 12 - 81782720 - 81783032 [+] on Build GRCm38 |
