siRNA Details
Gene Symbol: 4930423D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015188.1 | - | 2 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714739.1 | - | 3 | 292 | Chr. 10 - 117177629 - 117181114 [+] on Build GRCm38 |
