siRNA Details
Gene Symbol: 4930579H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016323.1 | - | 4 | 346 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715962.1 | - | 4 | 346 | Chr. 10 - 27389866 - 27396480 [+] on Build GRCm38 |
