siRNA Details
Gene Symbol: Gm12977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016328.1 | - | 1 | 42 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715967.1 | - | 1 | 42 | Chr. 4 - 133871830 - 133876482 [+] on Build GRCm38 |
