siRNA Details
Gene Symbol: Rap1gapos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007899.1 | - | 2 | 364 | ||
BQ287036.1 | - | 1 | 212 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707902.1 | - | 2 | 364 | Chr. 4 - 137696977 - 137706606 [-] on Build GRCm38 |
