siRNA Details
Gene Symbol: 2900019E01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013555.1 | - | 1 | 1438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF472917.1 | - | 1 | 300 | Chr. 15 - 43532662 - 43533204 [+] on Build GRCm38 |
