siRNA Details
Gene Symbol: 2900019A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013553.1 | - | 1 | 739 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV151516.1 | - | 1 | 234 | Chr. 5 - 24903368 - 24903656 [-] on Build GRCm38 |
