siRNA Details
Gene Symbol: 2900018K06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013551.1 | - | 1 | 76 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712516.1 | - | 1 | 74 | Chr. 2 - 136120255 - 136120721 [-] on Build GRCm38 |
