siRNA Details
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134899.1 | - | 1 | 1362 | ||
AK172916.1 | - | 19 | 5102 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134899.1 | - | 1 | 1362 | ||
AK172916.1 | - | 19 | 5102 |