siRNA Details
Gene Symbol: 1700023G08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006276.1 | - | 1 | 469 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706357.1 | - | 1 | 469 | Chr. 7 - 134261315 - 134261882 [-] on Build GRCm38 |
