siRNA Details
Gene Symbol: 1700023F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038039.1 | 3 | 268 | |||
GenBank mRNA | AK006272.1 | - | 3 | 269 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700023F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038039.1 | 3 | 268 | |||
GenBank mRNA | AK006272.1 | - | 3 | 269 |