siRNA Details
Gene Symbol: A530010L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079929.1 | - | 1 | 98 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724719.1 | - | 1 | 98 | Chr. 8 - 122761064 - 122762761 [-] on Build GRCm38 |
