siRNA Details
Gene Symbol: A530006G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046014.1 | 2 | 209 | |||
GenBank mRNA | AK079921.1 | - | 2 | 210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB636516.1 | - | 2 | 210 | Chr. 2 - 147710693 - 147716947 [+] on Build GRCm38 |
