siRNA Details
Gene Symbol: 5330417P21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019897.1 | - | 1 | 349 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716982.1 | - | 1 | 349 | Chr. 4 - 109069795 - 109070174 [-] on Build GRCm38 |
