siRNA Details
Gene Symbol: 2310039F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009692.1 | - | 1 | 141 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA734338.1 | - | 1 | 101 | Chr. 6 - 112758236 - 112758557 [-] on Build GRCm38 |
