siRNA Details
Gene Symbol: C030005H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021047.1 | - | 1 | 237 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C030005H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021047.1 | - | 1 | 237 |