siRNA Details
Gene Symbol: C030006F08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021049.1 | - | 1 | 152 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728110.1 | - | 1 | 152 | Chr. 6 - 97619679 - 97620295 [+] on Build GRCm38 |
