siRNA Details
Gene Symbol: A930037G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020939.1 | - | 1 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742361.1 | - | 1 | 422 | Chr. 5 - 22781340 - 22782009 [+] on Build GRCm38 |
