siRNA Details
Gene Symbol: 1700061H18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006853.1 | - | 2 | 442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706839.1 | - | 2 | 444 | Chr. 18 - 69943318 - 69969609 [-] on Build GRCm38 |
