siRNA Details
Gene Symbol: 1700061I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038029.1 | 8 | 1568 | |||
GenBank mRNA | AK006854.1 | - | 8 | 1570 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700061I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038029.1 | 8 | 1568 | |||
GenBank mRNA | AK006854.1 | - | 8 | 1570 |