siRNA Details
Gene Symbol: Gm16537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140856.1 | - | 98 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ061985.1 | - | 1 | 98 | Chr. 15 - 99715587 - 99724474 [-] on Build GRCm38 |
