siRNA Details
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086423.1 | - | 1 | 1732 | ||
AK142756.1 | - | 1 | 1732 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB525582.2 | - | 1 | 257 | Chr. 3 - 79641611 - 79642259 [-] on Build GRCm38 |
