siRNA Details
Gene Symbol: Gm816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136795.1 | - | 2 | 401 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721223.1 | - | 2 | 396 | Chr. 1 - 76892244 - 77027788 [-] on Build GRCm38 |
