siRNA Details
Gene Symbol: Gm11419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131774.1 | - | 2 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707204.1 | - | 2 | 318 | Chr. 11 - 81985406 - 81988769 [+] on Build GRCm38 |
