siRNA Details
Gene Symbol: 4933404K08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046038.1 | 3 | 432 | |||
GenBank mRNA | AK016652.1 | - | 3 | 433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716339.1 | - | 3 | 433 | Chr. 13 - 23278350 - 23281701 [+] on Build GRCm38 |
