siRNA Details
Gene Symbol: 4933438B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040685.1 | 1 | 146 | |||
GenBank mRNA | AK017106.1 | - | 1 | 153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716785.1 | - | 1 | 147 | Chr. 5 - 127030789 - 127084595 [+] on Build GRCm38 |
