siRNA Details
Gene Symbol: A330072L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039611.1 | - | 4 | 970 | ||
AK079662.1 | - | 4 | 970 |
Gene Symbol: Pydc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK165868.1 | - | 3 | 1866 |
