siRNA Details
Gene Symbol: Gm33180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139643.1 | - | 2 | 265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY725493.1 | - | 2 | 262 | Chr. 9 - 59500160 - 59512863 [-] on Build GRCm38 |
