siRNA Details
Gene Symbol: 4930555B11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040633.1 | 2, 3 | 266 | |||
GenBank mRNA | AK016127.1 | - | 2, 3 | 267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715729.1 | - | 2, 3 | 267 | Chr. 2 - 57181831 - 57187688 [+] on Build GRCm38 |
