siRNA Details
Gene Symbol: 2900072D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013764.1 | - | 1 | 952 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV154918.1 | - | 1 | 251 | Chr. 4 - 96030461 - 96030744 [+] on Build GRCm38 |
