siRNA Details
Gene Symbol: 2900072G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013767.1 | - | 1 | 687 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV154952.1 | - | 1 | 275 | Chr. 15 - 92350880 - 92351167 [+] on Build GRCm38 |
