siRNA Details
Gene Symbol: 2900072G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013766.1 | - | 1 | 1358 | ||
AW123148.1 | - | 1 | 216 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV068989.1 | - | 315 | Chr. - - [-] on Build |
