siRNA Details
Gene Symbol: 4930545H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045357.1 | 3 | 635 | |||
GenBank mRNA | AK016046.1 | - | 3 | 636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715634.1 | - | 3 | 632 | Chr. 10 - 68319079 - 68321139 [-] on Build GRCm38 |
