siRNA Details
Gene Symbol: 4930545E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045374.1 | 1 | 75 | |||
GenBank mRNA | AK016045.1 | - | 1 | 76 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715633.1 | - | 1 | 76 | Chr. 18 - 17568782 - 17580990 [-] on Build GRCm38 |
