siRNA Details
Gene Symbol: Gm12374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029761.1 | - | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615591.1 | - | 4 | 392 | Chr. 4 - 37249403 - 37320412 [+] on Build GRCm38 |
