siRNA Details
Gene Symbol: 5930427J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020038.1 | - | 1 | 1226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB099815.1 | - | 1 | 144 | Chr. 18 - 58091077 - 58091400 [-] on Build GRCm38 |
