siRNA Details
Gene Symbol: Gm38411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040387.1 | - | 763 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB209010.2 | - | 2 | 438 | Chr. 3 - 94368615 - 94370159 [-] on Build GRCm38 |
