siRNA Details
Gene Symbol: Gm38411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040387.1 | - | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635927.1 | - | 2 | 395 | Chr. 3 - 94369977 - 94377245 [-] on Build GRCm38 |
