siRNA Details
Gene Symbol: 1500002J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005119.1 | - | 1 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705314.1 | - | 1 | 500 | Chr. 8 - 85701072 - 85701831 [+] on Build GRCm38 |
