siRNA Details
Gene Symbol: Slc35a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009919.1 | - | 1 | 849 | ||
AK033430.1 | - | 1 | 2990 | |||
AK081236.1 | - | 1 | 2540 | |||
AK150620.1 | - | 1 | 1360 | |||
AK153354.1 | - | 1 | 1360 |
