siRNA Details
Gene Symbol: 2010110K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008384.1 | - | 119 | |||
AK029153.1 | - | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB613631.1 | - | 1 | 221 | Chr. 18 - 34751809 - 34754901 [+] on Build GRCm38 |
