siRNA Details
Gene Symbol: 2900012M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013530.1 | - | 1 | 1249 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY741722.1 | - | 1 | 249 | Chr. 9 - 10788902 - 10789556 [-] on Build GRCm38 |
