siRNA Details
Gene Symbol: 5730410E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017540.1 | - | 1 | 572 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717324.1 | - | 1 | 572 | Chr. 2 - 71357092 - 71357992 [-] on Build GRCm38 |
