siRNA Details
Gene Symbol: 4933436E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040455.1 | 3 | 256 | |||
GenBank mRNA | AK017079.1 | - | 3 | 256 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716757.1 | - | 3 | 252 | Chr. 1 - 139379456 - 139402670 [+] on Build GRCm38 |
