siRNA Details
Gene Symbol: 4933435N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017073.1 | - | 1 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716752.1 | - | 1 | 503 | Chr. 2 - 102922923 - 102923740 [-] on Build GRCm38 |
