siRNA Details
Gene Symbol: 4930432F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019591.1 | - | 1 | 1972 | ||
BC016220.1 | - | 1 | 997 | 4485816 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930432F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019591.1 | - | 1 | 1972 | ||
BC016220.1 | - | 1 | 997 | 4485816 |