siRNA Details
Gene Symbol: 1700060C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036606.2 | 1 | 12 | |||
GenBank mRNA | AK006839.1 | - | 1 | 13 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706821.1 | - | 1 | 13 | Chr. 2 - 158192008 - 158195744 [+] on Build GRCm38 |
