siRNA Details
Gene Symbol: 6530409C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018330.1 | - | 2 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719233.1 | - | 2 | 380 | Chr. 6 - 28215577 - 28218225 [+] on Build GRCm38 |
