siRNA Details
Gene Symbol: 4921513H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014881.1 | - | 1 | 1959 | ||
BC049716.1 | - | 2 | 446 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU961429.1 | - | 2 | 449 | Chr. 6 - 127776098 - 127778668 [-] on Build GRCm38 |
